DWK Life Sciences (Kimble) Blue Polypropylene Cap, 10/Cs
Representative image only.
DWK Life Sciences (Kimble) Blue Polypropylene Cap, 10/Cs
DWK Life Sciences (Kimble) | Mfr: 14395C-45| Item: 1596173
Mix, store, and transport culture media, chemicals, and solvents
₹2,600.00INR / Case of 10
₹2,600.00INR / Case of 10
Product Description
More About this Item
These heavy-duty general-purpose Type 1 borosilicate glass bottles have permanent white enamel graduations and marking areas. Each bottle includes an autoclavable (140°C/284°F) linerless blue polypropylene (PP) cap and drip-free pour ring. Maximum temperature: 450°C (840°F). Autoclavable.