DWK Life Sciences (Kimble) 953906-0090 ULTRA-WARE® Pickup Adapter, 90 mm with PTFE Tubing; 1/Cs
Representative image only.
DWK Life Sciences (Kimble) 953906-0090 ULTRA-WARE® Pickup Adapter, 90 mm with PTFE Tubing; 1/Cs
DWK Life Sciences (Kimble) | Mfr: 953906-0090| Item: 1595886
Modular system to filter HPLC mobile phase directly into reservoir
Call For Price
Call For Price
Product Description
Includes
Supplied with filtration cap, solvent pickup adapter and conical bottom reservoir
Key Features
  • Designed to filter HPLC mobile phase directly into the reservoir using 47mm membrane filter
  • Optional Conversion base allows use of 90mm membrane filter
  • In-line filtration with solvent pickup adapter eliminates traditional pour-and-wait filtration
  • Vacuum degassing is accomplished by closing the HI-VAC® valve
  • Patent No. 5,397,467
  • Recommended for most solvents used for HPLC mobile phase
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
  • Description90 mm Std. Pickup Adapter