Drierite 27069 RF 68 Refill kit for Gas Purifier, item number 01418-50
Representative image only.
Drierite 27069 RF 68 Refill kit for Gas Purifier, item number 01418-50
Drierite | Mfr: 27069 RF 68| Item: 0141860
250 mL of Drierite® desiccant and 250 mL of 5 Å molecular sieve for low-cost gas purifiers
₹14,800.00INR / Each
₹14,800.00INR / Each
Buy It Together
Current Item
Drierite 27069 RF 68 Refill kit for Gas Purifier, item number 01418-50
Drierite 27069 RF 68 Refill kit for Gas Purifier, item number 01418-50
0141860
₹14,800.00INR / Each
+ +
Drierite 12005 Nonindicating Desiccant,  6 Mesh; 5 lb Jar, 4/CS
0893413
₹54,500.00INR / Case of 4
=
Total Price 102700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Second half of column low-cost gas purifiers remove trace impurities
  • 1 year warranty
More About this Item
Purifier allows a maximum flow rate of 300 liters/hour and a maximum working pressure of 100 psi. Absorbs up to 25 g of moisture. Acrylic column has two halves—one half is filled with 250 ml of indicating Drierite desiccant, which produces a terminal dryness of 0.005 mg/liter. The other half is filled with 250 ml of 5 Å molecular sieves to remove trace impurities. Purifier measures 11-3/8"L x 2-5/8" dia and has an anodized aluminum cap, 303 stainless steel desiccant supports and coil springs, and an O-ring gasket. Inlet/outlet ports are 316 stainless steel; 1/8" threaded male connections.
1 Year Warranty
Specifications
  • Compatible WithItem number 01418-50
  • DescriptionRefill kit for Gas Purifier, item number 01418-50