Drierite 26960 Indicating Dry-Top Refillable Desiccant Stopper for 24/40 Neck Flasks

Representative image only.

Drierite 26960 Indicating Dry-Top Refillable Desiccant Stopper for 24/40 Neck Flasks

Drierite | Mfr: 26960| Item: 0893442

Keep reagents and samples dry in flasks

₹17,700.00INR / Each

₹17,700.00INR / Each

Buy It Together

Current Item

Drierite 26960 Indicating Dry-Top Refillable Desiccant Stopper for 24/40 Neck Flasks
Drierite 26960 Indicating Dry-Top Refillable Desiccant Stopper for 24/40 Neck Flasks
0893442

₹17,700.00INR / Each
+
Drierite 24001 Indicating Desiccant, 10-20 Mesh; 1 lb Jar, 12/CS
0893424

₹90,900.00INR / Case of 12
=
Total Price 108600.00 ADD 2 ITEMS TO CART
Product Description

Includes

Indicating desiccant (6.5 g).

Key Features

  • Fits standard 24/40 neck flasks
  • Translucent material allows for easy monitoring of desiccant color change
  • Chemically and thermally resistant
  • Easily refill indicating desiccant via screw cap
  • Stopper can be cleaned with detergent or solvent

More About this Item

This custom dry-top stopper with refillable inner chamber housing indicating desiccant, fits round-bottom and Erlenmeyer flasks with 24/40 necks. It offers you temporary or long-term storage in an anhydrous environment without requiring a desiccator. Composed of FEP (fluorinated ethylene propylene) plastic, the translucent stopper allows you to easily monitor color changes (from blue to pink) for constant visual assurance of active indicating desiccant. A vapor-permeable and chemically resistant membrane ensures the desiccant will never contaminate your sample.

Specifications
  • MaterialFluorinated Ethylene Propylene (FEP)
  • ColorTranslucent
  • DescriptionIndicating Dry-Top Refillable Desiccant Stopper for 24/40 Neck Flasks
  • Mesh Size10-20
Reviews
Q&A