Drager Tubes, Mercury, 0.05 to 2 mg/m3, 10/box
Representative image only.
Drager Tubes, Mercury, 0.05 to 2 mg/m3, 10/box
Drager | Mfr: CH23101| Item: 8651462
Reliable method for gas measurement
₹37,900.00INR / Box of 10
₹37,900.00INR / Box of 10
Buy It Together
Current Item
Drager Tubes, Mercury, 0.05 to 2 mg/m3, 10/box
Drager Tubes, Mercury, 0.05 to 2 mg/m3, 10/box
8651462
₹37,900.00INR / Box of 10
+
Drager CMS Analyzer Ammonia, 2 to 50 part per million
8651310
₹36,900.00INR / Each
=
Total Price 74800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Wider diameter tubes with well-spaced graduation marks are easier to read
  • Results are directly shown at the end of the measurement process
  • Fast measurement time—view results in 5 seconds to 15 minutes
  • 2 year warranty
More About this Item
Use gas detection tubes with the accuro® gas detector pump to measure various gases. Manually operated bellows pump draws a calibrated 100-mL sample through the short-term Dräger gas detection tubes to provide fast, accurate, reliable results.
Two year warranty
Specifications
  • Gas MeasuredMercury
  • Range0.05 to 2 mg/m³
  • TLV (ppm)0.025
  • DescriptionTubes, Mercury, 0.05 to 2 mg/m3, 10/box