Drager Diffusion Tube Nitrogen Dioxide 10/a-D
Representative image only.
Drager Diffusion Tube Nitrogen Dioxide 10/a-D
Drager | Mfr: 8101111| Item: 8651561
Direct read tubes identify workers exposure to chemicals
₹44,700.00INR / Box of 10
₹44,700.00INR / Box of 10
Product Description
Key Features
  • Read exposure levels easily but he markings on the side of the tube
  • 2 year warranty
More About this Item
Direct-reading diffusion tubes are used specifically for personal exposure monitoring. These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems. Exposure levels are easily read by the markings on the side of the tube.
Two year warranty
Specifications
  • Gas MeasuredNitrogen dioxide 10/a-D
  • Range1 hr: 10 to 200 ppm, 4 hr: 1.3 to 25 ppm
  • DescriptionDiffusion Tube Nitrogen Dioxide 10/a-D