Drager CMS Analyzer Ozone, 25 to 1000 ppb
Representative image only.
Drager CMS Analyzer Ozone, 25 to 1000 ppb
Drager | Mfr: 6406430| Item: 8651334
Eliminate user error for a consistent measurement every time
₹41,500.00INR / Each
₹41,500.00INR / Each
Buy It Together
Current Item
Drager CMS Analyzer Ozone, 25 to 1000 ppb
Drager CMS Analyzer Ozone, 25 to 1000 ppb
8651334
₹41,500.00INR / Each
+
Drager Tubes, Hydrogen Sulfide, 0 to 200 ppm, 10/box
8651438
₹33,100.00INR / Box of 10
=
Total Price 74600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital display of gas concentration eliminates ambiguous results
  • Internal recorder stores up to 50 measurements with time/date stamp
  • 2 year warranty
More About this Item
Just load a CMS gas specific chip into the bottom port of the analyzer. Easy-to-use, three-step slide switch allows analyzer to take measurement. Gas concentration is displayed in seconds with digital readout. Operates on four AA batteries (included). Chips contain 10 tests each and are sold separately.
Two year warranty
Specifications
  • Gas MeasuredOzone
  • Range25 to 100 ppb
  • DescriptionOzone, 25 to 1000 ppb