Diversey Virex® All Purpose Disinfectant Cleaner, 32 oz Spray Bottles; 8/Case
Representative image only.
Diversey Virex® All Purpose Disinfectant Cleaner, 32 oz Spray Bottles; 8/Case
Diversey | Mfr: CBD540533| Item: 7890107
Cleans and deodorizes all in one step
₹12,400.00INR / Case of 8
₹12,400.00INR / Case of 8
Product Description
Includes
Sprayers.
Key Features
  • Disinfects surfaces in three minutes
  • Kills Norovirus, VRE, and MRSA
  • Ready-to-use one-step disinfectant saves you time
  • Bactericide, tuberculocide, virucide, and fungicide sanitizer
  • Good for cleaning acrylic and polycarbonate barriers
More About this Item
Virex® all-purpose, quaternary-based disinfectant cleaner and deodorizer is ready to use out of the bottle – no diluting needed. Easily apply the cleaner with a mop, sponge, or cloth to hard, inanimate, nonporous surfaces such as floors, walls, porcelain, and plastic surfaces. Ideal for use in hospitals, nursing homes, schools, colleges, and hotels.
Meets OSHA’s bloodborne pathogen standards for decontaminating blood and body fluids. Effective in killing a broad spectrum of microorganisms including HBV, HIV-1, VRE, MRSA, MRSE, VISA, PRSP, Herpes Simplex types 1 & 2, Influenza Type A2, Adenovirus, Rotavirus, and Norovirus.
Also kills organisms that cause odors and works as an odor counteractant with a fresh citrus scent.
Specifications
  • FormatClear, liquid
  • Volume (Liters)0.95
  • DilutionReady to use
  • ApplicationsBactericide, tuberculocide, virucide, and fungicide sanitizer
  • DescriptionAll Purpose Disinfectant Cleaner; Case of eight 32 oz bottles with sprayers