Diversey Suma® Foam Free D9.6 General Degreaser, 32 oz Bottle; 12/CS
Representative image only.
Diversey Suma® Foam Free D9.6 General Degreaser, 32 oz Bottle; 12/CS
Diversey | Mfr: 903529270| Item: 7890160
Ready-to-use cleaner effectively cleans greasy and carbonized soils
₹33,800.00INR / Case of 12
₹33,800.00INR / Case of 12
Product Description
Includes
Foaming sprayer.
Key Features
  • Thick foam clings to surfaces for fast and easy cleaning without hard scrubbing
  • No fumes normally associated with harsh oven cleaners
  • pH of 13
More About this Item
Use this thick foam liquid cleaner to clean ovens, grills, hoods, and deep fryers. Simply spray directly on to surfaces, wait for several minutes, then then scrub the surface.
Do not use this product on aluminum or other soft metals, resilient floors, painted surfaces, or plastics. Do not use on continuous or self-cleaning ovens, or on surfaces above 180°F.
Available for sale to the 48 continental United States only.
Specifications
  • FormatClear amber, liquid
  • Volume (Liters)0.95
  • DilutionReady to use
  • ApplicationsClean greasy and carbonized soils
  • DescriptionSuma® Foam Free D9.6 General Degreaser, 32 oz; 12/CS