Diversey Suma® Dilac® D5.1 Acid-Based Lime Scale Remover, 32 oz Bottle; 12/CS
Representative image only.
Diversey Suma® Dilac® D5.1 Acid-Based Lime Scale Remover, 32 oz Bottle; 12/CS
Diversey | Mfr: 95325533| Item: 7890163
Effectively remove lime scale from stainless steel, soft metals, and plated metals
₹18,700.00INR / Case of 12
₹18,700.00INR / Case of 12
Product Description
Includes
Sprayer.
Key Features
  • Helps remove oxidation films to brighten dull metal surfaces
  • pH of 1.43
More About this Item
Acid-based lime scale remover is simple to use. Spray onto the surface and let it sit for about 15 minutes. Scrub the surface to brush away any lime scale then rinse.
Available for sale to the 48 continental United States only.
Specifications
  • FormatLight brown, liquid
  • Volume (Liters)0.95
  • DilutionReady to use
  • ApplicationsFor removing lime scales from stainless steel, plastics, and soft metals
  • DescriptionSuma® Dilac® D5.1 Acid-Based Lime Scale Remover, 32 oz Bottle; 12/CS