Diversey Oxivir® Tb Disinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea
Representative image only.
Diversey Oxivir® Tb Disinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea
Diversey | Mfr: 5627427| Item: 7890102
Tough on pathogens, not on people
₹65,300.00INR / Case of 4
₹65,300.00INR / Case of 4
Buy It Together
Current Item
Diversey Oxivir® Tb Disinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea
Diversey Oxivir® Tb Disinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea
7890102
₹65,300.00INR / Case of 4
+ =
Total Price 65300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Disinfects surfaces in less than one minute
  • Ready-to-use one-step disinfectant saves you time
  • Non-irritating to skin and eyes
  • Gentle formula suitable for daily use
  • Penetrates through the toughest soils
  • Kills SARS-CoV-2 (COVID-19 virus)
  • Choose liquid solution or perforated wipes
More About this Item
Oxivir® Tb is a one-step disinfectant cleaner that is ready to use out of the bottle—no diluting needed. Simply wipe any hard, nonporous surface and allow the surface to remain wet for one minute. With no added phosphates, phenols, and fragrance, Oxivir Tb cleaner meets the lowest EPA toxicity category and is environmentally responsible – active ingredients break down into oxygen and water. Formulated with Accelerated Hydrogen Peroxide technology to deliver fast-acting, broad spectrum disinfection with enhanced cleaning power.
Disinfectant is effective against key pathogens such as TB, MSRA, Norovirus, and VRE. It kills HBV, HCV, and HIV in one minute; and is a potent tuberculocide and fungicide to effectively clean, disinfect, and deodorize.
Note: Food contact surfaces must be rinsed with potable water after using. Do not use on glassware, utensils, or dishes. If you are unsure of compatibility, it is recommended to test on a small, discreet surface.
FOR COMMERCIAL AND INDUSTRIAL USE ONLY, no residential addresses accepted.
Specifications
  • FormatWipes
  • Width (InchShort)11
  • Height (InchShort)12
  • DilutionReady to use
  • ApplicationsGeneral virucide, bactericide, tuberculocide, fungicide
  • DescriptionDisinfectant Wipes, 11" x 12"; Case of 4 Canisters, 160 Wipes Ea