Diversey Oxivir® Tb Disinfectant Based Surface Wipes; 160 Wipes/Canister, 12 Canister/Case
Representative image only.
Diversey Oxivir® Tb Disinfectant Based Surface Wipes; 160 Wipes/Canister, 12 Canister/Case
Diversey | Mfr: 6292471| Item: 1639318
Oxivir Tb Wipes/ RTU Spray range are ready-to-use disinfectant cleaners, powered by AHP® technology, that disinfect hard nonporous surfaces in just 60 seconds
₹77,600.00INR / Case of 12
₹77,600.00INR / Case of 12
Product Description
Key Features
  • Kills coronavirus in 15 seconds
  • One-step cleaner and disinfectant Powered by Accelerated Hydrogen Peroxide technology
  • Realistic 60-second contact time increases the likelihood of proper usage and compliance
  • Declared by US EPA for products which can be used against SARS-CoV-2
  • No VOCs, no added fragrance and no NPEs
  • Environmentally responsible active ingredients break down to oxygen and water after use
  • Non-irritating to eyes and skin
  • Best in class HMIS rating 0-0-0
  • Lowest Hazard level in all six EPA toxicity categories
  • Available in RTU and Pre saturated Wipe format
More About this Item
Benefits:
• Effective against key pathogens TB, MRSA, Norovirus and VRE
• 1 minute kill for HBV, HCV, HIV-1 (AIDS virus), Tuberculocidal kill in 5 minutes and fungi in 10 minutes
Description:
Proprietary AHP technology deliver fast and effective cleaning performance, acting as a General Virucide, Bactericide, Tuberculocide, Fungicide, Sanitizer. Clean and disinfect high touch surfaces in just ONE Minute. Best in class HMIS rating and Lowest hazard level in EPA toxicity categories, not only make Oxivir TB safe to use around patients, visitors but also environmentally safe with end products of Oxygen and water.
Specifications
  • DescriptionDiversey Oxivir® Tb Disinfectant Based Surface Wipes; 160 wipes/Canister, 12 canister/Cs