Diversey Oxivir® RTU Surface Spray; 500 mL; 20/Case
Representative image only.
Diversey Oxivir® RTU Surface Spray; 500 mL; 20/Case
Diversey | Mfr: 6289802| Item: 1639317
A ready-to-use cleaner based on proprietary AHP technology
₹5,600.00INR / Case of 20
₹5,600.00INR / Case of 20
Product Description
Key Features
  • One-step , ready-to-use cleaner and powered by Accelerated Hydrogen Peroxide technology
  • Declared by US EPA for products which can be used against SARS-CoV-2
  • Spray format to ensure access to hard to reach areas
  • No VOCs, no added fragrance and no NPEs
  • Environmentally responsible active ingredients break down to oxygen and water after use
  • Non-irritating to eyes and skin
  • In use concentration of constituents, are nonhazardous to health per OSHA standards
More About this Item
Benefits:
• Effective against key pathogens TB, MRSA, Norovirus and VRE
• 1 minute kill for HBV, HCV, HIV-1 (AIDS virus); Tuberculocidal kill in 5 minutes; and fungi in 10 minutes
Description:
Proprietary AHP technology delivers fast, effective cleaning performance acting as a General Virucide, Bactericide, Tuberculocide, Fungicide, Sanitizer. Clean and disinfect high touch surfaces in just one minute. Safe for use around patients, by visitors people and on most surfaces.
Specifications
  • DescriptionDiversey Oxivir® RTU Surface Spray; 500 mL; 20/Case