Diversey Hygienizer Kit; includes RTU spray, dry wipes, hand sanitizer; 12 Kits/Case
Representative image only.
Diversey Hygienizer Kit; includes RTU spray, dry wipes, hand sanitizer; 12 Kits/Case
Diversey | Mfr: 6289992| Item: 1639319
A thoughtfully designed kit that helps you maintain personal hygiene with ease
₹10,000.00INR / Case of 12
₹10,000.00INR / Case of 12
Product Description
Key Features
  • Oxivir® RTU Liquid:
  • - Next generation ready to use single step disinfectant liquid cleaner based on proprietary AHP technology
  • - Declared by US EPA for products which can be used against SARS-CoV-2
  • - Nonhazardous to health per OSHA standards
  • Soft Care® Rinse-Free hand sanitizer:
  • - 70% alcohol based hand sanitizer
More About this Item
Benefits of RTU Spray:
• Effective against key pathogens TB, MRSA, Norovirus and VRE 1 minute kill for HBV, HCV, HIV-1 (AIDS virus); Tuberculocidal kill in 5 minutes; and fungi in 10 minutes
Description:
The Kit has been designed to assist each individual to maintain personal hygiene and ensure sanitization in their designated seating areas/cubicles in addition to the cleaning procedures carried out every day by the office housekeeping staff. The kit is a step in helping to counter the threat of COVID-19 lurking around us, providing high levels of sanitization in the most effective manner.
Kit Includes:
• 1 X 500 mL spray bottle with trigger Oxivir RTU One-Step Cleaner
• 1 X 200 mL bottle Soft Care Rinse-Free hand sanitizer
• 20X25 cm (25no) Dry Wipes; 100% polyester
Precautions:
1.For external use only.
2. Not for food contact surfaces
3. Product is not intended to be used on porous surfaces and computer peripherals
4. Direct inhalation of vapours should be avoided. 5. Don't spray Oxivir RTU on skin
Specifications
  • DescriptionDiversey Hygienizer Kit; includes RTU spray cleaner, hand sanitizer, dry wipes; 12 Kits/Case