Diversey Divoflow™ TC 86 Low-Foam Chlorinated Detergent for Hard Water; 55 Gallon Drum
Representative image only.
Diversey Divoflow™ TC 86 Low-Foam Chlorinated Detergent for Hard Water; 55 Gallon Drum
Diversey | Mfr: 57760010| Item: 7890147
Removes most organic materials
Call For Price
Call For Price
Product Description
Key Features
  • Free rising, minimizes water usage
  • Ideal for use in hard water
  • Minimizes acid cleaning cycles
More About this Item
This single-stage cleaner is a low foaming, chlorinated alkaline detergent/disinfectant ideal for use with clean-in-place (CIP) and spray washing equipment. Applications include the brewing, beverage, food, and dairy industries. Provides excellent removal of organic materials such as food soil, stains, and off-flavors for use in hard water conditions. Liquid can be fed and controlled automatically in CIP systems. It is free rinsing, leaves no hard water films, and minimizes acid cleaning cycles in most CIP operations.
Specifications
  • FormatClear light yellow liquid
  • Volume (gallons)55
  • DilutionReady to use
  • ApplicationsBrewing, beverage, food, and dairy
  • DescriptionLow Foam Chlorinated Detergent for Hard Water