Diversey Clax® Magic Rust Spot Remover, 32 oz Bottle; 6/CS
Representative image only.
Diversey Clax® Magic Rust Spot Remover, 32 oz Bottle; 6/CS
Diversey | Mfr: 100905320| Item: 7890161
Ready-to-use spot remover easily removes rust and iron stains
₹55,200.00INR / Case of 6
₹55,200.00INR / Case of 6
Product Description
Key Features
  • Convenient for use in commercial and on premise laundries
  • Apply directly to stain prior to washing
  • pH of 0.9
More About this Item
Special blend of solvents and other cleaning ingredients effectively remove discoloration caused by rust or iron stains. Simply saturate the stained area with the spot remover, gently scrape, let it sit for 5 to 10 minutes, then rinse. Ready-to-use solution is highly concentrated to limit the impact on the environment.
Available for sale to the 48 continental United States only.
Specifications
  • FormatClear, liquid
  • Volume (Liters)0.95
  • DilutionReady to use
  • ApplicationsRemoves rust and iron stains
  • DescriptionClax® Magic Rust Spot Remover, 32 oz Bottle; 6/CS