Distillation Apparatus Cap, Three Hole Delivery, 45 mm
Representative image only.
Distillation Apparatus Cap, Three Hole Delivery, 45 mm
| Item: 3462360
₹112,000.00INR / Case of 1
₹112,000.00INR / Case of 1
Product Description
More About this Item
This cap has three holes with 1/4"-28 threads—ideal for use with 1/8" OD tubing. Cap includes locknuts that allow positioning of PTFE tubing at any depth.
Specifications
  • DescriptionDistillation Apparatus Cap, Three Hole Delivery, 45 mm