Distillation Apparatus Cap, Open Top, without Liner, 45 mm
Representative image only.
Distillation Apparatus Cap, Open Top, without Liner, 45 mm
| Item: 3462314
₹59,500.00INR / Case of 10
₹59,500.00INR / Case of 10
Product Description
More About this Item
PBTP caps without liner. Ideal for use when temperatures are between 49° to 356°F(-45 to 180°C). Open top caps may be used with liners sold separately, or may be used with syringes for reagent introduction or sampling.
Specifications
  • DescriptionDistillation Apparatus Cap, Open Top, without Liner, 45 mm