Digi-Sense Type-K, 2-Wire Feedthrough, 1 / 2" NPT (M) Fitting, 48" PTFE Wire
Representative image only.
Digi-Sense Type-K, 2-Wire Feedthrough, 1 / 2" NPT (M) Fitting, 48" PTFE Wire
Digi-Sense | Item: 9387002
₹19,600.00INR / Each
₹19,600.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Use these two-wire feedthroughs to make connections inside vessels or chambers. Use with probes with miniconnectors. Feedthroughs withstand vacuum conditions or pressures to 100 psi. Stainless steel and epoxy construction. Temperature range: -40 to 250°> F (-40 to 121°> C). Feedthroughs have a 1/2'' NPT(M) fitting. Include 48''L, PTFE insulated, 24-gauge (0.0201'' wire dia) lead wires.
6 Month Warranty
Specifications
  • Connector TypeType-K
  • MaterialPTFE
  • Wire Gauge (Ga)24
  • Min Temperature (° F)-40
  • Max Temperature (° F)250
  • Min Temperature (° C)-40
  • Max Temperature (° C)121
  • DescriptionType-K, 2-Wire Feedthrough, 1 / 2" NPT (M) Fitting, 48" PTFE Wire