Digi-Sense TI.ST Dual-Magnet Surface Thermometers; Range 50 - 750F
Representative image only.
Digi-Sense TI.ST Dual-Magnet Surface Thermometers; Range 50 - 750F
Digi-Sense | Item: 0810705
Inexpensive way to easily measure surface temperature
₹10,700.00INR / Each
₹10,700.00INR / Each
Product Description
Key Features
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
  • 1 year warranty
More About this Item
An ideal choice for use on ovens, boilers, process lines, motors, or anywhere a surface temperature needs to be measured. The specially designed bimetal sensing element and housing provide quick readings. These thermometers feature a dual magnet mount that will attach to an ferrous surface, steel case, glass lens and polished aluminum 2" dial with easy-to-read graduations.
1 Year Warranty
Specifications
  • Min Temperature (° F)50
  • Max Temperature (° F)750
  • Division (° F)10
  • Dial Size (in)2
  • Dial / Case Material304 Stainless Steel
  • Stem Material304 Stainless Steel
  • DescriptionDual-Magnet Surface Thermometers; Range 50 - 750F