Digi-Sense Thermocouple Mounting Panel, Horizontal, Mini Connectors; 6 Circuits
Representative image only.
Digi-Sense Thermocouple Mounting Panel, Horizontal, Mini Connectors; 6 Circuits
Digi-Sense | Item: 1852737
Anodized aluminum construction for full corrosion protection
₹10,600.00INR / Each
₹10,600.00INR / Each
Product Description
Key Features
  • Create easy access to temperature connections in control rooms, test stands, laboratories, and plants
  • Complete your panel assembly with insert panel thermocouple connectors
  • Available in 2, 4, 5 and 6 circuits for vertical face plates and 6, 8, 10 and 12 circuits for horizontal face plates
  • 6 month warranty
More About this Item
Use to creat easy to access direct temperature connection in control rooms, test stands, laboratories, and plants. Use with insert panel temperature connectors to create a complete panel assembly. Available in 2, 4, 5 & 6 circuits for Vertical Face Plates and 6, 8, 10 & 12 circuits for Horizontal Face Plates.
6 Month Warranty
Specifications
  • Connector TypeMini connector
  • Special InformationAnodized aluminum face plate/mounting panel
  • Length (in)4 29/64
  • Height (in)1 1/2
  • DescriptionThermocouple Mounting Panel, Horizontal, Mini Connectors; 6 Circuits