Digi-Sense Standard Connector/Cable Clamp
Representative image only.
Digi-Sense Standard Connector/Cable Clamp
Digi-Sense | Item: 1852789
Protect connections against vibrations and tugging
₹960.00INR / Each
₹960.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire clamps secure and stabilize thermocouple and RTD wire/connector junctions. Use clamps where physical stresses lead to failed junctions. Available in both standard and miniconnector sizes.
6 Month Warranty
Specifications
  • DescriptionStandard Connector/Cable Clamp