Digi-Sense RTD Connector, White 3-Blade ANSI, Male, 5/Pk
Representative image only.
Digi-Sense RTD Connector, White 3-Blade ANSI, Male, 5/Pk
Digi-Sense | Item: 1852785
Easily connect RTD probes to a handheld meter
₹7,500.00INR / Pkg of 5
₹7,500.00INR / Pkg of 5
Product Description
Key Features
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Screw-top connection
  • 6 month warranty
More About this Item
Choose between white three-blade ANSI miniconnectors or round 3-pin connectors (Oakton® and Digi-Sense® meters only). Available individually and in packs of five.
6 Month Warranty
Specifications
  • Connector TypePlug
  • ColorWhite
  • Special InformationGlass-filled nylon body
  • Max Temperature (° F)300
  • Max Temperature (° C)149
  • DescriptionRTD Connector, White 3-Blade ANSI, Male, 5/Pk