Digi-Sense RTD Connector, 3 Pin, Female, 5/Pk
Representative image only.
Digi-Sense RTD Connector, 3 Pin, Female, 5/Pk
Digi-Sense | Item: 1852783
Easily connect RTD probes to a handheld meter
₹33,400.00INR / Pkg of 5
₹33,400.00INR / Pkg of 5
Product Description
Key Features
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • Connection requires soldering
  • 6 month warranty
More About this Item
Choose between white three-blade ANSI miniconnectors or round 3-pin connectors (Oakton® and Digi-Sense® meters only). Available individually and in packs of five.
6 Month Warranty
Specifications
  • Connector TypeJack
  • Special InformationNickel-plated metal & black plastic
  • Max Temperature (° F)300
  • Max Temperature (° C)149
  • DescriptionRTD Connector, 3 Pin, Female, 5/Pk