Digi-Sense RT24-3-527 RTD Wire, 24-Gauge, Tfe Insulation, 100-ft Spool
Representative image only.
Digi-Sense RT24-3-527 RTD Wire, 24-Gauge, Tfe Insulation, 100-ft Spool
Digi-Sense | Item: 0854231
Extend the measuring distance of your probes
₹38,600.00INR / Each
₹38,600.00INR / Each
Product Description
Key Features
  • 100-foot spool of RTD wire- cut to any lenght needed!
  • 6 month warranty
More About this Item
Three-strand RTD wire features excellent resistance to abrasion, chemicals, and moisture. Each silver-plated copper strad is color-coded with fluorinated ethylene propylene (FEP). The three starnds are twisted to increase flexibility and minimize electromagnetic noise, then jacketed in white FEP. The FEP insulation provides temperature resistance up to 400°F, and chemical resistance to solvents, acids, and oils. Additionally, the insulation protects the wire from thermal aging, while maintaining its strength and flexibility.
6 Month Warranty
Specifications
  • MaterialPTFE
  • Wire Gauge (Ga)24
  • Inner InsulationPolytetrafluoroethylene
  • Outer InsulationPolytetrafluoroethylene
  • Stranded / SolidStranded
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)100
  • DescriptionRTD Wire, 24-Gauge, Tfe Insulation, 100-ft Spool