Digi-Sense Oven Verification Thermometer; 50 to 200C, PFA Coated
Representative image only.
Digi-Sense Oven Verification Thermometer; 50 to 200C, PFA Coated
Digi-Sense | Item: 9030073
Enclosed bottle ensures accuracy and is ideal for use in environmental chambers
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Digi-Sense Oven Verification Thermometer; 50 to 200C, PFA Coated
Digi-Sense Oven Verification Thermometer; 50 to 200C, PFA Coated
9030073
Call For Price
+ + =
Total Price 54800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • An individually serial number is printed on the thermometer stem
  • Nonmercury blue spirit liquid
  • NIST- traceable statement of accuracy included
  • 5 year warranty
More About this Item
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. These thermometers are supplied in a media-filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
5 Year Warranty
Specifications
  • Length (")5 45/64
  • Min Temperature (° C)50
  • Max Temperature (° C)200
  • Accuracy±2.5°C; ±5.0°C above 100°C
  • Division (° C)2
  • Thermometer FillBlue Spirit
  • Bottle FillVermiculite
  • Overall Length (InchShort)5.7
  • Overall Length (mm)145
  • Stem MaterialPFA-Coated White-Back Glass
  • DescriptionOven Verification Thermometer; 50 to 200C, PFA Coated
  • Warranty5 years