Digi-Sense Locking Miniconnector, Type-K Thermocouple, Male, 1/Ea
Representative image only.
Digi-Sense Locking Miniconnector, Type-K Thermocouple, Male, 1/Ea
Digi-Sense | Item: 1852752
No more unwanted disconnections
₹2,000.00INR / Each
₹2,000.00INR / Each
Product Description
Key Features
  • Self-locking plugs and jacks secure connection against vibrations and tugging
  • Glass-filled nylon construction provides rigidity, strength, and surface hardness
  • 6 month warranty
More About this Item
These plugs and jacks feature a locking mechanism designed to prevent inadvertent uncoupling. Each flat-blade style connector is ANSI color-coded by thermocouple type: black (J), yellow (K), or blue (T).
6 Month Warranty
Specifications
  • Connector TypePlug
  • Special InformationGlass-filled nylon body
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • DescriptionLocking Miniconnector, Type-K Thermocouple, Male, 1/Ea