Digi-Sense Liquid-In-Glass Refrigerator/Freezer Thermometer; -40 to 27C (-40 to 80F), Steel Case
Representative image only.
Digi-Sense Liquid-In-Glass Refrigerator/Freezer Thermometer; -40 to 27C (-40 to 80F), Steel Case
Digi-Sense | Item: 9025060
Call For Price
Call For Price
Product Description
Key Features
  • For reference only (should not be calibrated)
  • 1 year warranty
More About this Item
This low-temperature thermometer is ideal for refrigerators and freezers. Thermometer features a dual-scale range of -40 to 80°F (-40 to 27°C) with divisions of 2°F (1°C). The large numbers and magnified lens make reading easy. Red spirit-filled thermometer is made of plastic and includes stainless steel clips to easily mount and hang from shelves.
1 Year Warranty
Specifications
  • Min Temperature (° F)-40
  • Length (")4 29/32
  • Min Temperature (° C)-40
  • Max Temperature (° F)80
  • Max Temperature (° C)27
  • Accuracy±2.0°F (±1.0°C)
  • Division (° C)1
  • Division (° F)2
  • Thermometer FillRed Spirit
  • Dial / Case MaterialSteel
  • Overall Length (InchShort)4.9
  • Overall Length (mm)124
  • Stem MaterialGlass
  • DescriptionLiquid-In-Glass Refrigerator/Freezer Thermometer; -40 to 27C (-40 to 80F), Steel Case