Digi-Sense Dual Thermocouple Connector, Mini, Female, Type-K; 5/pk
Representative image only.
Digi-Sense Dual Thermocouple Connector, Mini, Female, Type-K; 5/pk
Digi-Sense | Item: 1852777
Captive cover screw for easy assembly
₹14,200.00INR / Pkg of 5
₹14,200.00INR / Pkg of 5
Product Description
Key Features
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • 6 month warranty
More About this Item
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
6 Month Warranty
Specifications
  • Connector TypeK
  • Special InformationGlass filled nylon body
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • DescriptionDual Thermocouple Connector, Mini, Female, Type-K; 5/pk