Dickson A793 Fire Hydrant Adapter Kit, Hydrant to 1/4" NPT
Representative image only.
Dickson A793 Fire Hydrant Adapter Kit, Hydrant to 1/4" NPT
Dickson | Mfr: A793| Item: 0841086
Make the connections you need for your data logger
₹71,300.00INR / Each
₹71,300.00INR / Each
Product Description
Key Features
  • Special fitting and adapters allow you to connect our pressure units to almost any source
More About this Item
Kits contain multiple fittings and adapters to easily connect your pressure measuring devices to almost any source.
This chrome plated adapter converts the opening of a fire hydrant to a 1/4" NPT connector.
It is compatible with Dickson PW4 units, PR8 units, PR100, PR300, PR500, PR125, PR325, and PR525.
Specifications
  • Set IncludesHydrant to 1/4" NPT adapter
  • DescriptionFire Hydrant Adapter Kit, Hydrant to 1/4" NPT