Dickson A792 Steam Pressure Pigtail for PW4/PW8 Chart Recorders
Representative image only.
Dickson A792 Steam Pressure Pigtail for PW4/PW8 Chart Recorders
Dickson | Mfr: A792| Item: 6851161
Protect your equipment from the effects of live steam
₹18,000.00INR / Each
₹18,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This pigtail steam siphon is used when monitoring the pressure of steam. Filling the pigtail with water acts as a barrier between the steam and the sensor.
The Pressure Pigtail is compatible with PW4 units, PR8 units, PR100, PR300, PR500, PR125, PR325, and PR525.
1 Year Warranty
Specifications
  • MaterialBrass
  • Connections1/4" x 1/4" NPT(M)
  • DescriptionSteam Pressure Pigtail for PW4/PW8 Chart Recorders