Dickson A721 Locking Case for PR125/325/525 Loggers
Representative image only.
Dickson A721 Locking Case for PR125/325/525 Loggers
Dickson | Mfr: A721| Item: 6851135
Monitor pressure up to 500psi at-a-glance or with one-step data retrieval
₹34,200.00INR / Each
₹34,200.00INR / Each
Product Description
Key Features
  • USB-enabled triple speed downloading
  • 1-second sampling rate
  • IP68 case
  • FLASH card option
  • User replaceable battery
  • Pressure sensor includes built in diaphragm seal
  • 1 year warranty
More About this Item
One-step data retrieval from datalogger to a FLASH memory card makes the routine of gathering data fast and easy. By pressing a single button, all your data will be downloaded to the removable memory card (optional), thereby eliminating the need to bring a laptop or move the datalogger. A Card Reader Kit, 23036-52, and a memory card, 23036-53, are all you need for this data retrieval system.
Choose from 0 to 100 psi, 0-300 psi, or 0 to 500 psi models. All models feature LCD display with min/max option, unit of measure indicator, low battery indicator, and much more. An optional locking case is available to prevent tampering.
1 Year Warranty
Specifications
  • DescriptionLocking Case for PR125/325/525 Loggers