Dickson A203 6" High-Temperature Probe for Circular Temperature Chart Recorder
Representative image only.
Dickson A203 6" High-Temperature Probe for Circular Temperature Chart Recorder
Dickson | Mfr: A203| Item: 0851606
Connects to temperature chart recorder that accepts type K thermocouple probes
₹56,200.00INR / Each
₹56,200.00INR / Each
Product Description
Key Features
  • Measures temperatures up to 1093°C (2000°F), tip only
  • Made of 316 stainless steel
  • High-temperature probe works with Dickson circular chart recorder
  • 6 month warranty
More About this Item
Temperature probe to accurately measure high temperatures up to 1093°C (2000°F). Easily connect to a chart recorder that accepts a type K miniconnector for permanent record of your measurements.
6 Month Warranty
Specifications
  • ConnectionsType K miniconnector
  • Description6" High-Temperature Probe for Circular Temperature Chart Recorder
  • Warranty6 months