Dibafit™ Multiport Connector, 3-Way, 1/4"-28 UNF(F) flat bottom, PEEK™, mountable; 1/ea
Representative image only.
Dibafit™ Multiport Connector, 3-Way, 1/4"-28 UNF(F) flat bottom, PEEK™, mountable; 1/ea
Diba | Mfr: 001057| Item: 2193911
Use with Omnifit® gripper fittings or Omni-Lok™ Type P/S for an inert fluid path with zero dead volume
₹10,400.00INR / Each
₹10,400.00INR / Each
Product Description
Key Features
  • Constructed of durable PEEK
More About this Item
These block connectors have 1/4"-28 UNF(F) threads and 1.5 mm internal bore. Select from 3-way or 4-way configuration.
Specifications
  • Length (")1 1/16
  • Fitting MaterialPEEK
  • Width (InchShort)1 1/16
  • Height (InchShort)1/2
  • Number Of Ports3
  • Thread size UNF(F) (port 2) (InchShort)1/4"-28
  • Thread size UNF(F) (port 3) (InchShort)1/4"-28
  • Thread size UNF(F) (port 1) (InchShort)1/4"-28
  • DescriptionDibafit™ Multiport Connector, 3-Way, 1/4"-28 UNF(F) flat bottom, PEEK™, mountable; 1/ea