Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, 1/4"-28 UNF x 1/16" OD; 10/Pk
Representative image only.
Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, 1/4"-28 UNF x 1/16" OD; 10/Pk
Diba | Mfr: 2310| Item: 2194000
Chemically inert flangeless fitting with permanently attached PTFE ferrule ideal for repeat connect/disconnect
₹15,100.00INR / Pkg of 10
₹15,100.00INR / Pkg of 10
Product Description
Key Features
  • Available in PTFE/SS or PTFE/PEEK material
More About this Item
The Dibafit™ Gripper fitting system uses a one-piece ferrule which fits permanently onto the outside of semi-rigid tubing.
Specifications
  • Fitting MaterialPTFE
  • Connection Typeflat bottom, 1/4"-28
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Max Operating Pressure (PSI)1000
  • DescriptionGripper Ferrule, 1/4"-28 UNF flat bottom, 1/16" OD tubing; 10/pk