Diba Omnifit® Q-Series Solvent Bottle Cap, GL38/38-430 (plastic), 2 UNF(F) Ports w/ Valves, Blue
Representative image only.
Diba Omnifit® Q-Series Solvent Bottle Cap, GL38/38-430 (plastic), 2 UNF(F) Ports w/ Valves, Blue
Diba | Mfr: 00938PQ-2V| Item: 2194282
On/off valves control individual fluid lines
₹32,400.00INR / Each
₹32,400.00INR / Each
Buy It Together
Current Item
Diba Omnifit® Q-Series Solvent Bottle Cap, GL38/38-430 (plastic), 2 UNF(F) Ports w/ Valves, Blue
Diba Omnifit® Q-Series Solvent Bottle Cap, GL38/38-430 (plastic), 2 UNF(F) Ports w/ Valves, Blue
2194282
₹32,400.00INR / Each
+ =
Total Price 44100.00 ADD 2 ITEMS TO CART
Product Description
Includes
A 12-1/4" (31 cm) long, 1/8" OD PTFE tubing
Key Features
  • Robust threaded ports accept 1/4"-28 UNF(M) fittings
  • Anti-twist design
  • Caps 21942-82 and -83 are designed to fit plastic bottles
  • Designed to fit plastic bottles.
More About this Item
These Q-series solvent caps ensure a leak-free, vapor-tight seal to protect lab staff from exposure to toxic fumes, prevent ingress of particulates, and prevent evaporation of solvent. Use the on/off valves to control the flow of each line independently. The anti-twist design lets the PTFE body spin freely within the cap. No need to disconnect the fluid lines before unscrewing the cap—no more twisted tubes when the cap is fitted or removed.
Easily connect semi-rigid tubing to caps using 1/4"-28 UNF(M) fittings. Connect flexible tubing using Dibafit™ PEEK™ thread-to-barb adapters. Each port features a stainless steel thread insert to enable repeated connect/disconnect. All caps are autoclavable. A 12-1/4” (31 cm) long, 1/8" OD PTFE tubing is supplied fitted inside the cap and extending into the bottle.
Cap is designed to fit plastic bottles. Plastic bottles are unsuitable for vacuum or pressurized applications.
Design for 38-430 thread is different for glass and plastic bottles. Caps 21942-82 and -83 are designed to fit plastic bottles which are unsuitable for vacuum or pressurized applications.
Specifications
  • Cap MaterialPP outer cap, PTFE body, and PCTFE internal rotor
  • Cap / Stopper Size38-430 (GL38)
  • Cap ColorBlue
  • AutoclavableYes
  • Special Features2 x 1/4"-28 UNF(F)
  • DescriptionDiba Omnifit® Q-Series Solvent Waste Cap, GL38/38-430 (plastic), 2 UNF(F) ports w/ valves, blue; 1/ea