Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/8" (3.2 mm); 4/pk
Representative image only.
Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/8" (3.2 mm); 4/pk
Diba | Mfr: 001504| Item: 2193948
Select the size you need for a leaktight seal
₹4,500.00INR / Pkg of 4
₹4,500.00INR / Pkg of 4
Product Description
Key Features
  • Chemically resistant PTFE construction
More About this Item
Select PTFE cones when chemical resistance is important. The OD of the tubing should be matched to the inside diameter of the cone.
Specifications
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.2
  • DescriptionOmnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1/8" (3.2 mm); 4/pk