Diba Omnifit® Filter Bubbler, 316 Stainless Steel; 1/ea
Representative image only.
Diba Omnifit® Filter Bubbler, 316 Stainless Steel; 1/ea
Diba | Mfr: 009SFB| Item: 2194342
Push-fit onto a 1/8" OD tube for a quick connection
₹11,800.00INR / Each
₹11,800.00INR / Each
Product Description
Key Features
  • Optimize solvent filtration with filters and bubblers
More About this Item
These reservoir filters and bubbler provide simple and effective solvent filtration. A 10 µm filter ensures even the smallest particulates are removed from the solvent supply line. Bottom-of-bottle filters have slots in the body allowing virtually all liquid to be removed from the bottle, minimizing waste. Select from PTFE body/membrane or PTFE body/PE membrane filters. Filter bubbler, suitable for filtration or sparging applications, comes with a 316 SS filter.
Specifications
  • Membrane Material316 Stainless steel
  • Width (InchShort)1/2
  • Pore Size (micron)10 µm
  • Height (InchShort)1 3/8
  • Housing MaterialPTFE
  • ConnectionsFor any bottle
  • Ports1/8" OD tube connection
  • DescriptionDiba Omnifit® Filter Bubbler, 316 Stainless Steel; 1/ea