Diba Omnifit® EZ & EZ SolventPlus Packing Sleeve, 3 mm; 1/ea

Representative image only.

Diba Omnifit® EZ & EZ SolventPlus Packing Sleeve, 3 mm; 1/ea

Diba | Mfr: 006PS-03| Item: 1194557

Simplify column filling and packing

₹9,500.00INR / Each

₹9,500.00INR / Each
Product Description

Key Features

  • Join 2 columns or connect to column packer with 3 mm inside diameter

More About this Item

Use these packing sleeves to join two columns for filling and packing.

Specifications
  • Column Inside Diameter (ID) (mm)3
Reviews
Q&A