Diba Omnifit® Bottom-of-Bottle Filter for GL45 Bottle, 10 µm PTFE; 1/ea
Representative image only.
Diba Omnifit® Bottom-of-Bottle Filter for GL45 Bottle, 10 µm PTFE; 1/ea
Diba | Mfr: 009F-45| Item: 2194334
Push-fit onto a 1/8" OD tube for a quick connection
₹11,200.00INR / Each
₹11,200.00INR / Each
Product Description
Key Features
  • Optimize solvent filtration with filters and bubblers
More About this Item
These reservoir filters and bubbler provide simple and effective solvent filtration. A 10 µm filter ensures even the smallest particulates are removed from the solvent supply line. Bottom-of-bottle filters have slots in the body allowing virtually all liquid to be removed from the bottle, minimizing waste. Select from PTFE body/membrane or PTFE body/PE membrane filters. Filter bubbler, suitable for filtration or sparging applications, comes with a 316 SS filter.
Specifications
  • Membrane MaterialPTFE
  • Pore Size (micron)10 µm
  • Housing MaterialPTFE
  • ConnectionsFor GL45 bottle
  • Ports1/8" OD tube connection