Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk
Representative image only.
Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk
Diba | Mfr: 008FK47| Item: 2194056
Fits permanently onto the outside of semi-rigid tubing
₹20,900.00INR / Pkg of 10
₹20,900.00INR / Pkg of 10
Buy It Together
Current Item
Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk
Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk
2194056
₹20,900.00INR / Pkg of 10
+
Diba Omni-Lok™ Assembly Tool for Type-S Fittings; 1/ea
2194042
₹112,000.00INR / Each
=
Total Price 132900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Rated for pressures up to 500 psi (34 bar)
More About this Item
Type S system uses a two-piece ferrule made up of an inner PEEK™ or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
Specifications
  • Fitting MaterialPEEK ferrule/SS locking ring
  • Compression Size / Tube Od (Port 1) (in)3/16
  • DescriptionType-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk