Diba Omni-Lok™ Type-P Ferrule, 3/16" OD tubing, 1/4"-28 flat bottom & M6, PTFE / SS; 10/pk
Representative image only.
Diba Omni-Lok™ Type-P Ferrule, 3/16" OD tubing, 1/4"-28 flat bottom & M6, PTFE / SS; 10/pk
Diba | Mfr: 008FT47| Item: 2194059
Fits permanently onto the outside of semi-rigid tubing
₹30,800.00INR / Pkg of 10
₹30,800.00INR / Pkg of 10
Buy It Together
Current Item
Diba Omni-Lok™ Type-P Ferrule, 3/16" OD tubing, 1/4"-28 flat bottom & M6, PTFE / SS; 10/pk
Diba Omni-Lok™ Type-P Ferrule, 3/16" OD tubing, 1/4"-28 flat bottom & M6, PTFE / SS; 10/pk
2194059
₹30,800.00INR / Pkg of 10
+ =
Total Price 45700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rated for pressures up to 1000 psi (69 bar)
  • Use with flat-bottom 1/4"-28 UNF(M) or M6 ISO(M) fittings
More About this Item
Type P system uses a one-piece ferrule with only the inner PTFE sealing face in the fluid path. Select from PTFE/SS or PTFE/PEEK™ material of construction.
Specifications
  • Fitting MaterialPTFE/SS
  • Compression Size / Tube Od (Port 1) (in)3/16
  • DescriptionType-P Ferrule, 3/16" OD tubing, 5/16"-24 flat bottom, PTFE / SS; 10/pk