Diba Omni-Lok™ Tubing End Fitting, Type S/P, 1/16"OD tubing, 6-40 UNF(M), PEEK; 10/pk
Representative image only.
Diba Omni-Lok™ Tubing End Fitting, Type S/P, 1/16"OD tubing, 6-40 UNF(M), PEEK; 10/pk
Diba | Mfr: 008NF16-KC9L| Item: 2194200
Permanently attached ferrules are ideal for repeated connect/disconnect
₹15,800.00INR / Pkg of 10
₹15,800.00INR / Pkg of 10
Buy It Together
Current Item
Diba Omni-Lok™ Tubing End Fitting, Type S/P, 1/16"OD tubing, 6-40 UNF(M), PEEK; 10/pk
Diba Omni-Lok™ Tubing End Fitting, Type S/P, 1/16"OD tubing, 6-40 UNF(M), PEEK; 10/pk
2194200
₹15,800.00INR / Pkg of 10
+ =
Total Price 31400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Free spinning fitting eliminates tubing from twisting
  • A recess in the fitting houses the ferrule allowing maximum thread engagement with the port
More About this Item
These fittings use a two-piece ferrule that fits permanently to the outside of the tubing. Select from PEEK ferrules with a tapered port or with flat port and stainless steel lock ring.
Specifications
  • Fitting MaterialPEEK
  • Length (")19/64
  • Width (InchShort)1/8
  • Connection TypeCompression x UNF(M)
  • Number Of Ports2
  • Thread size UNF(M) (port 1) (InchShort)6-40
  • Compression Size / Tube Od (Port 2) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)500
  • DescriptionOmni-Lok™ Tubing End Fitting, Type S/P, 1/16"OD tubing, 6-40 UNF(M), PEEK; 10/pk