Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, 1/4"-28 UNF(M), green PP; 10/pk
Representative image only.
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, 1/4"-28 UNF(M), green PP; 10/pk
Diba | Mfr: 008NF32-YC5G| Item: 2194230
Permanently attached ferrules are ideal for repeated connect/disconnect
₹4,200.00INR / Pkg of 10
₹4,200.00INR / Pkg of 10
Buy It Together
Current Item
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, 1/4"-28 UNF(M), green PP; 10/pk
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, 1/4"-28 UNF(M), green PP; 10/pk
2194230
₹4,200.00INR / Pkg of 10
+ =
Total Price 14300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Free spinning fitting eliminates tubing from twisting
  • A recess in the fitting houses the ferrule allowing maximum thread engagement with the port
More About this Item
Type P ferrule has inner PTFE sealing face, the only material in the fluid path. Two-piece type S ferrule consists of an inner PEEK or ETFE sealing face surrounded by a stainless steel lock ring, with only the inner portion in the fluid path. Choose type P for higher pressures, type S for higher temperatures. Select glass-filled polypropylene compact head fittings available in a range of colors for easy identification. Long head fitting is ideal where accessibility is an issue. Use short thread option for most thread engagement in shallow ports.
Specifications
  • Length (")3/4
  • Fitting MaterialPolypropylene
  • Width (InchShort)1/4
  • Connection TypeCompression x UNF(M)
  • Number Of Ports2
  • Thread size UNF(M) (port 1) (InchShort)1/4"-28
  • Compression Size / Tube Od (Port 2) (in)1/8
  • ColorGreen
  • Max Operating Pressure (PSI)500
  • DescriptionOmni-Lok™ Compact Tubing End Fitting, Type S/P, 1/8"OD, 1/4"-28 UNF(M), green PP; 10/pk