Diba Column O-Ring, 6.6 mm, FKM; 10/pk
Representative image only.
Diba Column O-Ring, 6.6 mm, FKM; 10/pk
Diba | Mfr: 006OR-06-V| Item: 1194546
Select from FKM or FFKM (Chemraz®) O-rings
₹2,400.00INR / Pkg of 10
₹2,400.00INR / Pkg of 10
Product Description
Key Features
  • Enhance performance of you Diba Omnifit® chromatography columns
  • O-ring has 6.6 mm inner diameter
  • Constructed of FKM material
More About this Item
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.
Specifications
  • Column Inside Diameter (ID) (mm)6.6
  • Wetted MaterialsFKM