Diba Column O-Ring, 50 mm, FFKM; 2/pk
Representative image only.
Diba Column O-Ring, 50 mm, FFKM; 2/pk
Diba | Mfr: 006OR-50-C| Item: 1194555
Select from FKM or FFKM (Chemraz®) O-rings
₹106,000.00INR / Pkg of 2
₹106,000.00INR / Pkg of 2
Product Description
Key Features
  • Enhance performance of you Diba Omnifit® chromatography columns
  • O-ring has 50 mm inner diameter
  • Constructed of FFKM material
More About this Item
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.
Specifications
  • Column Inside Diameter (ID) (mm)50
  • Wetted MaterialsFFKM