Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16"OD tubing, 1/4"-28 UNFM, black PC; 10/pk
Representative image only.
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16"OD tubing, 1/4"-28 UNFM, black PC; 10/pk
Diba | Mfr: 008NC16-CS5B| Item: 2194162
Removable and reusable system for quick and convenient low-pressure connection
₹5,600.00INR / Pkg of 10
₹5,600.00INR / Pkg of 10
Buy It Together
Current Item
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16"OD tubing, 1/4"-28 UNFM, black PC; 10/pk
Diba Click-N-Seal™ Tubing End Fitting, inverted cone, 1/16"OD tubing, 1/4"-28 UNFM, black PC; 10/pk
2194162
₹5,600.00INR / Pkg of 10
+ =
Total Price 9800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • When the cap clicks, you know the seal is tight
  • Self-torquing fittings create a secure seal without the use of any tools
More About this Item
Use fittings with inverted cones to achieve a simple and easy-to-use low-pressure connection. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. The ETFE cone is the only part in the fluid path and offers excellent chemical resistance. The cone sits in the recessed area of the fitting allowing maximum thread engagement. None of the parts are permanently attached to the tubing, so both fittings and inverted cones can easily be removed and reused. Select from acetal, polycarbonate (PC), or PEEK™ material of construction. PEEK fittings are compact in design and provide excellent chemical resistance.
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)1/4-28
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.57
  • ColorBlack
  • Max Operating Pressure (PSI)250
  • DescriptionClick-N-Seal™ Tubing End Fitting, inverted cone, 1/16"OD tubing, 1/4"-28 UNFM, black PC; 10/pk