Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk
Representative image only.
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk
Diba | Mfr: 008NF16-KT9R| Item: 2194206
Hand tighten fittings without tools to create a secure seal
₹13,000.00INR / Pkg of 10
₹13,000.00INR / Pkg of 10
Buy It Together
Current Item
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk
2194206
₹13,000.00INR / Pkg of 10
+ =
Total Price 28600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • When the cap clicks, you know the seal is tight
More About this Item
These unique self-torquing fittings create a secure seal without the use of any tools. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. Fittings are made of PEEK™ and are compact in design and provide excellent chemical resistance.
The fitting can spin freely, eliminating tubing from twisting. A recess in the fitting houses the ferrule allowing maximum thread engagement with the port. Type S ferrule system uses a two-piece ferrule that fits permanently to the outside of the tubing. Two-part ferrule is made up of an inner PEEK or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
Specifications
  • Fitting MaterialPEEK
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (InchShort)6-40
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.57
  • ColorRed
  • DescriptionClick-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk