Delmhorst CX-30 Cotton Moisture Meter
Representative image only.
Delmhorst CX-30 Cotton Moisture Meter
Delmhorst | Mfr: CX-30W/CS| Item: 5980085
The CX-30 is essential for maintaining ideal moisture in cotton from harvest through ginning, storage, and selling, preventing equipment damage and preserving cotton quality and strength
₹121,000.00INR / Each
₹121,000.00INR / Each
Buy It Together
Current Item
Delmhorst CX-30 Cotton Moisture Meter
Delmhorst CX-30 Cotton Moisture Meter
5980085
₹121,000.00INR / Each
+
Delmhorst 30-E/C  Baled Fibers Electrode
5982302
₹26,400.00INR / Each
=
Total Price 147400.00 ADD 2 ITEMS TO CART
Product Description
Includes
Meter and carrying case.
Key Features
  • Accurate moisture ranges for lint cotton, seed cotton, wool, and viscose/rayon
  • Large, auto-backlit display with on-screen statistics for easy reading and tracking
  • Bluetooth® connectivity with the EDGE® app for data export, timestamping, and geotagging
  • Temperature correction and customizable alarms for user-set moisture levels
  • Ergonomic, rugged ABS case with E-Z grip and durable buttons
  • 2 year warranty
More About this Item
Trusted by cotton producers worldwide, the CX-30 Cotton Moisture Meter delivers reliable, precise moisture monitoring across all production phases. Designed for ease of use, the CX-30 features an intuitive display, internal calibration, and customizable alerts to prevent moisture issues that could damage equipment or reduce cotton value. Together with the Delmhorst EDGE® app, it offers seamless data export and location tagging, ensuring quality and durability from field to sale.
Specifications
  • Measurement TypeLint cotton (4-18%), seed cotton (6-20%), wool (12-30%), viscose/rayon (6-25%)
  • Temperature CompensationYes
  • Min Temperature (° F)0
  • Min Temperature (° C)-18
  • Max Temperature (° F)255
  • Max Temperature (° C)124
  • Display TypeLED
  • Data LoggingYes
  • Memory Capacity100 readings
  • AlarmUser-selected MC level
  • OutputBluetooth®
  • Battery2 AA
  • Electrode TypeNone included
  • Height (in)1 19/32
  • Width (in)2 27/32
  • Length (in)8 1/32
  • DescriptionCotton Moisture Meter
  • Warranty2 years