Delmhorst 608/4 21E Replacement Moisture Pins, 4" Long; 4/Pk

Representative image only.

Delmhorst 608/4 21E Replacement Moisture Pins, 4" Long; 4/Pk

Delmhorst | Mfr: 608/4| Item: 5982042

The wide assortment of replacement pins gives the flexibility to choose the right combination for your application

₹16,700.00INR / Each

₹16,700.00INR / Each
Product Description

Key Features

  • Configure your meter to your needs

More About this Item

Delmhorst offers the widest selections of electrodes and accessories in the industry. This allows you to customize you meter to meet your specific requirement, and each electrode is interchangeable with any Delmhorst moisture meter with a quick connection..

4 per pack. Replacement pin for the 21-E electrode

Specifications
  • Measurement TypePin
  • Length (in)4
  • Description21E Replacement Moisture Pins, 4" Long; 4/Pk
Reviews
Q&A